Learn More
SCAT™ Flat Flange Lids for Glass Reactors
Product Code. 17849056 Shop All SCAT Europe Products
Click to view available options
Includes:
1 x NPT 1/2 in. (f) Centered, 1 x NPT 1/2 in. (f) Directly in Center, 1 x NPT 1 in. (f) Directly in Center, 1 x NPT 1/2 in. (f) Beside Center, 1 x NPT 1/4 in. (f) Beside Center, 1 x NPT 1/4 in. (f) Vertical
1 x NPT 1/2 in. (f) Centered, 1 x NPT 1/4 in. (f) Directly in Center, 2 x NPT 1/2 in. (f) Beside Center
1 x NPT 1/2 in. (f) Centered, 2 x NPT 1/2 in. (f) Directly in Center, 1 x NPT 1/2 in. (f) Beside Center
1 x NPT 1/2 in. (f) Centered, 2 x NPT 1/2 in. (f) Directly in Center, 1 x NPT 1/2 in. (f) Beside Center, 1 x NPT 1/4 in. (f) Beside Center, 1 x NPT 1/4 in. (f) Vertical
1 x NPT 1/2 in. (f) Centered, 2 x NPT 1/2 in. (f) Directly in Center, 1 x NPT 1/2 in. (f) Beside Center, 1 x NPT 3/4 in. (f) Beside Center, 1 x NPT 1/4 in. (f) Vertical
1 x NPT 1/2 in. (f) Centered, 3 x NPT 1/2 in. (f) Beside Center
Joint Type:
DN 100
DN 150
DN 60
Unit Size:
Each
Product Code. 17849056
Brand: SCAT™ 130060
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
The flat flange lid is usually placed between two flanges to be joined and then tightened with bolts to create a secure connection. Suitable for glass reactors with a SCHOTT™ flat flange.
  • Flat flange allows the lid to be securely attached to the reactor body and ensures a reliable seal to prevent leakage
  • Compatible according to DIN 12214 (laboratory flat flange) and compatible with glass reactors for seamless integration
  • Effortless and hassle-free to assemble, saving time and effort
  • Flexibly expand our products to fit your specific needs and meet future needs
  • Made entirely of PTFE, for universal chemical resistance to a wide range of harsh chemicals, increasing application versatility and reliability
  • Equipped with practical attachments that allow easy connection and use of existing equipment that has ground or GL threads
  • Important component for the safe and reliable connection of pipes and vessels in various industrial applications to prevent leaks and keep equipment running smoothly
Spezifikation
Product Type Flat Flange
For Use With (Application) Connection of Pipes and Vessels, Preventing Leaks
For Use With (Equipment) Glass Reactors
Includes 1 x NPT 1/2 in. (f) Centered, 1 x NPT 1/4 in. (f) Directly in Center, 2 x NPT 1/2 in. (f) Beside Center
Joint Type DN 60
Material PTFE
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt