Learn More
Helios Italquartz Flat-Bottom Dishes, Translucent
Flat-bottom dishes constructed of translucent, durable, resistant, and chemically inert quartz glass.
Brand: Helios Italquartz 61F00149
Additional Details : Weight : 0.20000kg
Description
- Translucent
- Flat bottom
- Chemically inert
- Excellent electrical insulation
- Excellent strength and durability
- Exceptional thermal stability compared to glass
Specifications
Translucent | |
55 cc | |
Round | |
18 mm |
Flat | |
Quartz | |
Evaporating Dish | |
77 mm |