Learn More
Helios Italquartz Flat-Bottom Dishes, Translucent
Flat-bottom dishes constructed of translucent, durable, resistant, and chemically inert quartz glass.
Brand:  Helios Italquartz 61F00148
Additional Details : Weight : 0.20000kg
 View more versions of this product
Product Code. 15808014
  • 149.00€ / Each
Estimated Shipment: 13-06-2024
Description
  • Translucent
  • Flat bottom
  • Chemically inert
  • Excellent electrical insulation
  • Excellent strength and durability
  • Exceptional thermal stability compared to glass
Specifications
Specifications
Translucent
35 cc
Round
15 mm
Flat
Quartz
Evaporating Dish
68 mm