Learn More
JULABO™ Flat Bath Cover for PURA™ Water Baths
Product Code. 16216611
Click to view available options
For Use With (Equipment):
PURA™ 10
PURA™ 14
PURA™ 22
PURA™ 30
Type:
1 opening, 190 mm dia.
10 openings, 115 mm dia.
2 openings, 190 mm dia.
3 openings, 190 mm dia.
4 openings, 92 mm dia.
6 openings, 115 mm dia.
6 openings, 92 mm dia.
8 openings, 92 mm dia.
Unit Size:
Each
Product Code. 16216611
Brand: JULABO™ 9970573
Flat bath covers for PURA™ Water Baths, with appropriate set of rings
  • Accessory for PURA™ water bath
  • With appropriate sets of rings
Specifications
Description Flat bath cover
For Use With (Equipment) PURA™ 22
Type 8 openings, 92 mm dia.