Learn More
Fisherbrand™ Flat Base Gamma Radiation Shields
Product Code. 12317187 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Height (Metric):
600 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Unit Size
12317187 600 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12317187
Estimated Shipment: 26-02-2027
Fixed radiation shielding ideal for protection from gamma radiation.
  • Gamma radiation, shield, fixed 45 degree angle, flat base, dimensions 600h x 350w x 300d mm
  • Provides clear, non-distorted vision, reducing light images and shadows, especially when the user is in a sitting position
  • Available in two sizes, with either a flat base to provide bench protection, or a curved base which fits conveniently over safety trays
    • Supplied in small, medium and large sizes and with curved base 15cm-deep for use with safety trays or flat 30x30cm base for under-the-bench protection
    • A range of angles offers increased maneuverability, while clear optical acrylic aids visualization
Specifications
Product Type Gamma Shield
Width (Metric) 350 mm
Height (Metric) 600 mm
Thickness (Metric) 350 mm
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.