Learn More
Zeiss™ Fl Filter Set 09
Description
- For fluorescence applications
- Excitation: BP 450-490
- Beam Splitter: FT 510
- Emission: LP 515
Specifications
Specifications
| Type | FL Filter Set 09 |
| Quantity | 1 Ea. |