Learn More
Zeiss™ Fl Filter Set 09
Product Code. 11875810
Change view
Click to view available options
Type:
FL Filter Set 09
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
11875810 FL Filter Set 09 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11875810 Supplier Zeiss™ Supplier No. 4880099901000
Ensure optimum specimen preservation with these filter sets
  • For fluorescence applications
  • Excitation: BP 450-490
  • Beam Splitter: FT 510
  • Emission: LP 515
Specifications
Type FL Filter Set 09
Quantity 1 Ea.