Learn More
Zeiss™ FL Attenuator
Product Code. 11865480
Click to view available options
Type:
Slider FL Attenuator
Unit Size:
Each
Product Code. 11865480
Brand: Zeiss™ 4237309100000
This item requires a price update prior to being ordered.
Request a Price for this item here.
Insert this discrete attenuator into the aperture stop plane of a reflected-light illuminator. Filter wheel with 6 positions: Pos. 1: clear aperture; Pos. 2-6: transmission appr. 70, 50, 40, 20 and 2%
Compatible Microscopes:
  • Axio Examiner.D1
  • Axio Examiner.Z1
  • Axio Scope.A1
  • Axio Vert.A1 FL
  • Axio Vert.A1 MAT
Specifications
Type Slider FL Attenuator
For Use With (Application) For insert into the aperture stop plane of reflected-light illuminator
Includes Filter wheel with 6 positions
Quantity 1 Ea.
Size 14 x 40 mm