Learn More
Thermo Scientific™ Fixed-Needle, Super-Elastic Plunger Syringe
Product Code. 15204573 Shop All Thermo Scientific Products
Click to view available options
Volume (Metric):
10 μL
Unit Size:
Each
Product Code. 15204573
Brand: Thermo Scientific™ 365D5416
Thermo Scientific™ syringes for GC instruments are designed to provide enhanced durability,clarity and accuracy in sample introduction.
TRUSTED_SUSTAINABILITY
Specifications
Needle Gauge 23 to 26 G
Product Type GC Autosampler Syringe, Fixed Needle, Super-Elastic Plunger
Volume (Metric) 10 μL
Length (Metric) Needle 42 mm
Needle Point Style Cone
For Use With (Equipment) Agilent GC Instruments