Learn More
Thermo Scientific™ Removable-Needle Autosampler Syringes for GC Instruments
Product Code. p-4525916 Shop All Thermo Scientific Products
Change view
Click to view available options
Volume (Metric):
5 μL
10 μL
50 μL
100 μL
250 μL
Needle Gauge:
23 G
23 to 26s G
26 G
31 G
Quantity:
6 Pk.
Ea.
Needle Point Style:
Cone
Side Hole
Unit Size:
Each
Pack of 6
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Needle Gauge Quantity Needle Point Style unitSize
10177654 5 μL 23 G 6 Pk. Cone Pack of 6
10299254 5 μL 23 to 26s G Ea. Cone Each
10188044 5 μL 23 to 26s G 6 Pk. Cone Pack of 6
10641406 250 μL 26 G Ea. Cone Each
10658525 100 μL 23 G Ea. Cone Each
10350925 100 μL 23 G Ea. Side Hole Each
10793877 250 μL 23 G Ea. Side Hole Each
10524275 10 μL 31 G Ea. Cone Each
17864961 50 μL 23 G Ea. Cone Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 10177654 Supplier Thermo Scientific™ Supplier No. 365C0956
Only null left
Add to basket
Add to basket
Thermo Scientific™ Removable-Needle Autosampler Syringes for GC Instruments are accurate and reproducible over a range of volumes.
  • Borosilicate glass with metal flanges
  • Needles and plungers are stainless steel
  • Field-proven syringe design
TRUSTED_SUSTAINABILITY
Specifications
Material Stainless Steel (Needle), Stainless Steel (Plunger)
Description 23 gauge; 5uL; 42m length; 6/Pk.
Needle Gauge 23 G
Length (Metric) 42 mm
Product Type GC Autosampler Syringe, Fixed Needle
Disposable Reusable
Quantity 6 Pk.
Volume (Metric) 5 μL
Length (Metric) Needle 42 mm
Needle Point Style Cone
For Use With (Equipment) Agilent GC Instruments
Connection Type Fixed