Learn More
Thermo Scientific™ Removable-Needle Autosampler Syringes for GC Instruments
Product Code. 10299254 Shop All Thermo Scientific Products
Click to view available options
Volume (Metric):
5 μL
10 μL
50 μL
100 μL
250 μL
Needle Gauge:
23 G
23 to 26s G
26 G
31 G
Quantity:
6 Pk.
Ea.
Needle Point Style:
Cone
Side Hole
Unit Size:
Each
Pack of 6
Product Code. 10299254
missing translation for 'mfr': Thermo Scientific™ 365C0971
Thermo Scientific™ Removable-Needle Autosampler Syringes for GC Instruments are accurate and reproducible over a range of volumes.
  • Borosilicate glass with metal flanges
  • Needles and plungers are stainless steel
  • Field-proven syringe design
TRUSTED_SUSTAINABILITY
Specifications
Material Stainless Steel (Needle), Stainless Steel (Plunger)
Description 23 to 26 gauge; 5uL; 42mm length
Needle Gauge 23 to 26s G
Length (Metric) 42 mm
Product Type GC Autosampler Syringe, Fixed Needle
Disposable Reusable
Quantity Ea.
Volume (Metric) 5 μL
Length (Metric) Needle 42 mm
Needle Point Style Cone
For Use With (Equipment) Agilent GC Instruments
Connection Type Fixed