Learn More
Zeiss™ Fixed Analyzer
Click to view available options
Type:
Fixed Analyzer Slide D with Lambda Plate
Unit Size:
Each
Description
Compatible Microscopes:
- Axio Lab.A1
- Axio Lab.A1 FL-LED
- Axio Lab.A1 MAT
- Axio Lab.A1 Pol
- Axioskop 2
- Axioskop 2 MAT
- Axioskop 20
- Axioskop 40
- Axioskop 40 Pol
Specifications
Specifications
| Type | Fixed Analyzer Slide D with Lambda Plate |
| Color | Black |
| Dimensions (L x W) | 6 x 20 mm |
| Includes | Lambda Plate |
| Quantity | 1 Ea. |