Learn More
BRAND™ Fitting for Dr. Schilling Pattern Automatic Burette
Product Code. p-7159971 Shop All Brand Products
Click to view available options
For Use With (Equipment):
15 mL Dr. Schilling Pattern Automatic Burette
25 to 50 mL Dr. Schilling Pattern Automatic Burette
Unit Size:
Each
Product Code. 10340151
Brand: BRAND™ 23746
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Micrometer screw with press button, including support clamp
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 15 mL
For Use With (Equipment) 15 mL Dr. Schilling Pattern Automatic Burette
Includes Micrometer Screw with Press Button, Support Clamp
Type Buret Fitting