missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FITC Mouse, Clone: 3C10A1, Proteintech
Mouse Monoclonal Antibody
140.00€ - 399.00€
Specifications
Antigen | FITC |
---|---|
Clone | 3C10A1 |
Concentration | 1000 μg/mL |
Applications | ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17721199
|
Proteintech
68132-1-IG-20UL |
20 μL |
140.00€
20µL |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
17711199
|
Proteintech
68132-1-IG-150UL |
150 μL |
399.00€
150µL |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
Description
FITC Monoclonal antibody specifically detects FITC in fitc samples. It is validated for ELISASpecifications
FITC | |
1000 μg/mL | |
Monoclonal | |
Liquid | |
PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
Primary | |
Store at -20°C |
3C10A1 | |
ELISA | |
Unconjugated | |
Mouse | |
IgG2a | |
Protein A purification | |
Antibody |