Specifications
| Content And Storage | Store at -20 C. Avoid freeze-thaw cycles. |
| Product Line | Novus |
| Quantity | 1 mg |
| For Use With (Application) | Purple Solid. Soluble in DMSO (>10 mg/mL). |
| Target | FITC-geldanamycin |
| Type | Geldanamycin |
| Label or Dye | FITC |