Learn More
Fisherbrand™ Zipper Seal Sample Bags
Product Code. p-32048 Shop All Fisherbrand Products
Click to view available options
Dimensions (L x W):
18 x 9 in. (46 x 23cm)
3 x 2 in. (7 x 5cm)
8 x 5 in. (20 x 13cm)
Thickness (English):
2 mil
4 mil
Material:
LDPE
Polyethylene
Unit Size:
Pack of 100
Pack of 1000
Pack of 50
Product Code. 13554530
Zipper-locking feature securely seals bags
  • Durable, transparent polyethylene
  • Ideal for collecting, storing, and protecting laboratory specimens and samples easily and conveniently
  • Clear, print-free surface lets user mark and label each bag separately
Not suggested for use with liquids
Specifications
Autoclavable Not Autoclavable
Capacity (English) 5 oz.
Closure Type Zip Seal
Color Clear
Material LDPE
No. per Case 1000
No. per Pack 100
Thickness (English) 2 mil
Dimensions (L x W) 3 x 2 in. (7 x 5cm)
Length (English) 3 in.
Length (Metric) 7 cm
Width (English) 2 in.
Width (Metric) 5 cm