Learn More
Masterflex™ Tight-Seal™ Clamps
Product Code. 10527070 Shop All Fisherbrand Products
Click to view available options
Diameter (Metric) Inside:
0.6 to 0.7 cm
0.914 to 1.01 cm
1.04 to 1.16 cm
1.193 to 1.34 cm
1.34 to 1.52 cm
1.49 to 1.67 cm
1.701 to 1.981 cm
1.85 to 2.1 cm
2.41 to 2.76 cm
2.61 to 2.99 cm
3.17 to 3.6 cm
Diameter (English) Inside:
0.24 to 0.29 in.
0.36 to 0.4 in.
0.41 to 0.46 in.
0.47 to 0.53 in.
0.53 to 0.6 in.
0.59 to 0.66 in.
0.67 to 0.78 in.
0.73 to 0.83 in.
0.95 to 1.09 in.
1.03 to 1.18 in.
1.25 to 1.42 in.
Unit Size:
1 set
Each
Pack of 100
Product Code. 10527070
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
One-piece, reusable nylon clamps for hoses and tubing.
One-piece, reusable nylon clamps for hoses and tubing
  • Chemical and corrosion resistant
  • With wide temperature tolerance
  • Squeeze shut for leakproof seal, press on side to release
  • Gas or chemically sterilized for biomedical or food applications
  • 12 sizes accommodate tubing with an O.D. of 0.246 to 1.425 in. (6.25 to 36.19mm)
Pressure, vacuum, and liquid applications
Specifications
Quantity 100/Pk.
Type Hose and Tubing Clamp
Material Nylon
For Use With (Equipment) Hose and tubing with 0.24 to 1.42 in O.D.
Diameter (English) Inside 0.59 to 0.66 in.
Temperature (Metric) Resistant 0°C to 90°C
Temperature (English) Resistant 32°F to 194°F
Diameter (Metric) Inside 1.49 to 1.67 cm