Learn More
Fisherbrand™ Silicone Recessed Septum Stoppers
Product Code. 11542503 Shop All Fisherbrand Products
Click to view available options
Diameter (Metric) Inner:
13.75 to 14.25 mm
14.50 to 15.50 mm
20.00 to 21.00 mm
9.22 to 9.75 mm
Unit Size:
Pack of 10
Product Code. 11542503
Nontoxic and autoclavable recessed septum stoppers are externally serrated to create an effective internal seal
  • Elliptical flange to allow efficient removal of stopper from aperature
  • Easily pierced with a hypodermic needle
Specifications
Diameter (Metric) Inner 9.22 to 9.75 mm
For Use With (Application) Nontoxic and autoclavable recessed septum stoppers are externally serrated to create an effective internal seal
For Use With (Equipment) For 9.22 to 9.75mm I.D. containers
Color Translucent
Material Silicone
Product Type Septum Stopper