Learn More
Fisherbrand™ Round Stir Bars with Removable Pivot Ring
Product Code. p-317329 Shop All Fisherbrand Products
Click to view available options
Length (Metric):
25.4 mm
Diameter (Metric):
8 mm
Length (English):
1 in.
Diameter (English):
0.31 in.
Unit Size:
Each
Product Code. 15525604
Only null left
Add to basket
Add to basket
Stir bars have polygon multifaceted surfaces that add turbulence relative to similar smooth size cylindrical bars.
  • Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon™ PTFE low friction coating; durable and inert
  • Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
  • Autoclave safe
  • Small removable pivot ring in the center adds to their versatility
  • Pivot ring minimizes contact area of bar to vessel
  • Ring also reduces friction and the marring of plastic containers
  • No pivot ring is included on the two smallest sizes, Cat. Nos. 14-512-140 and -141
Specifications
Autoclavable Autoclavable
Certifications/Compliance ISO 9001:2008
Color White
Description PTFE Coated Alnico
Diameter (English) 0.31 in.
Length (English) 1 in.
Coating PTFE
Diameter (Metric) 8 mm
Length (Metric) 25.4 mm
Material Alnico Magnet, PTFE
Quantity 1
Shape Sphere
Type Stir Bar