Learn More
Hellma™ Quartz, Silica and Glass Standard Cell Cuvettes
Product Code. 11721774
Click to view available options
Path Length:
1 mm
5 mm
10 mm
Material:
Quartz Suprasil 300
Suprasil Quartz
near-UV Glass
Quantity:
2/Pk.
Ea.
Unit Size:
Each
Pack of 2
Product Code. 11721774
Brand: Hellma™ 1001020
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Includes
Fitted PTFE covers or tapered PTFE stoppers, except 14-385-906A and -902A, which come with glass cover plates
Designed to hold samples for spectroscopic experiments and optical analysis. Hellma™ Quartz, Silica and Glass Standard Cell Cuvettes provide optically excellent transparency.
  • Quartz - high UV transmission and low thermal expansion
  • Glass - used in the visible range of the spectrum
  • Low values of refractive index and dispertion
  • Good resistance to chemicals
Specifications
Material near-UV Glass
Quantity 2/Pk.
Path Length 10 mm