Learn More
Fisherbrand™ Powder Funnels
GREENER_CHOICE
Product Code. 11997914 Shop All Fisherbrand Products
Click to view available options
Diameter (Metric)Top:
6.5cm
7.5cm
10.5cm
15cm
Diameter (Metric) Stem:
16mm
21mm
27mm
Capacity (Metric):
50 mL
100 mL
225 mL
750 mL
Height (Metric):
6.6 cm
8.4 cm
10.3 cm
13.7 cm
Unit Size:
Pack of 24
Pack of 36
Pack of 4
Pack of 6
Product Code. 11997914
Only null left
Add to basket
Add to basket
Parallel stems minimize bridging of powder.
Parallel stems minimize bridging of powder
  • Durable polypropylene
  • External ribbing prevents air locks
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric)Top 6.5cm
Capacity (Metric) 50 mL
Diameter (Metric) Stem 16mm
Material Polypropylene
Height (Metric) 6.6 cm
Autoclavable Autoclavable
Length (Metric) Stem 22mm