Learn More
Fisherbrand™ Porcelain Crucible Covers
Product Code. 13419709 Shop All Fisherbrand Products
Click to view available options
Diameter (Metric) Outer:
30mm
40mm
44mm
53mm
60mm
75mm
87mm
99mm
115mm
Diameter (Metric) Inner:
26mm
34mm
38mm
45mm
54mm
66mm
77mm
90mm
105mm
Height (Metric):
5mm
5.5mm
6mm
7mm
8mm
9mm
Thickness (Metric):
1.75mm
1.9mm
2.3mm
2.5mm
2.75mm
3mm
Unit Size:
Pack of 6
Product Code. 13419709
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
For use with Fisherbrand High- and Wide-Form Porcelain Crucibles.
For use with Fisherbrand High- and Wide-Form Porcelain Crucibles
  • Glazed inside and out except for the outside bottom surface
  • With perforated handle
  • Strongly recommended that heating and cooling rates not exceed 200°C per hour and maximum use temperature of 1100°C
Specifications
Description With Perforated handle
Diameter (Metric) Inner 26mm
Material Porcelain
Finish Glazed
For Use With (Application) Use with Fisherbrand high and wide form porcelain crucible
Max. Temperature (Metric) 1150°C
Thickness (Metric) 1.75mm
Weight (Metric) 6g
Diameter (Metric) Outer 30mm
Height (Metric) 5mm
Product Type Crucible Cover