Learn More
Fisherbrand™ Flexible-Arm™ Replacement Light Bulb
Product Code. 15253976 Shop All Fisherbrand Products
Change view
Click to view available options
Wattage:
20 W
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Wattage unitSize
15253976 20 W Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15253976
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Replacement part for Flexible-Arm™ light
Replacement part for Flexible-Arm™ light
  • Halogen bulb
  • 20w
  • Soft-white light
Specifications
Light Source Halogen
Wattage 20 W
Color Soft White
For Use With (Equipment) Flexible-Arm light
Product Type Soft-white Light Bulb