Learn More
Fisherbrand™ Erlenmeyer Flask Clamps for Ultrasonic Units
Product Code. 10562963 Shop All Fisherbrand Products
Click to view available options
For Use With (Application):
1000mL erlenmeyer flask
250mL Erlenmeyer flask
50 mL Erlenmeyer Flask
500 mL Erlenmeyer Flask
Unit Size:
Each
Product Code. 10562963
Safely and securely attach Erlenmeyer flasks to Fisherbrand™ ultrasonic cleaning units. Fisherbrand™ Erlenmeyer Flask Clamps are manufactured using strong and durable stainless steel for superior stability.
  • Safe and secure attachment
  • Manufactured from strong and durable stainless steel
  • Provides superior stability
Specifications
Type Flask Clamp
For Use With (Equipment) Ultrasonic Cleaner
For Use With (Application) 1000mL erlenmeyer flask