Learn More
Fisherbrand™ Borosilicate Glass Disposable Serological Pipets with Regular Tip, Standard Length, Nonsterile
Product Code. p-3102145
Click to view available options
Volume (Metric):
1 mL
Graduations:
0.01 mL
Negative Graduations:
0.2 mL
Color:
Yellow
Unit Size:
Pack of 250
Product Code. 13267499
Brand: Fisherbrand™ 7212011100
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Long, slender, tapered tips speed pipetting
Regular-tipped pipets have long, slender, tapered tips to speed pipetting
Chemically inert pipets have negative graduations as well as bidirectional scales.
  • Blue markings are easy to read
  • Packaging meets requirements of small-, medium-, or large-volume use
  • Multipack with resealable flap for larger volume use permits easy access to several pipets at a time without contaminating unused pipets
  • Outer packaging is recyclable
Order Info
Bulk packed.
Compliance
Regular-tip styles have volume tolerances that comply with ASTM™ E 714. Calibrated To Deliver/Blow Out,
TRUSTED_SUSTAINABILITY
Specifications
Calibration To Deliver
Certifications/Compliance USP type I, ASTM E438
Material Borosilicate Glass
Sterility Non-sterile
Graduated Yes
Graduations 0.01 mL
Negative Graduations 0.2 mL
Tip Style Regular
Tolerance ±0.03 mL
Packaging Type Bulk
Volume (English) 0.03 oz.
Color Yellow
Volume (Metric) 1 mL
Type Serological Pipet