Learn More
Fisherbrand™ ASTM™ Specific Gravity Hydrometers for Liquids Lighter than Water
Product Code. 11819270 Shop All Fisherbrand Products
Click to view available options
ASTM Number:
103H
104H
105H
106H
107H
82H
83H
84H
85H
86H
87H
88H
Certifications/Compliance:
3-point Cert, ASTM E100-14 T1
3-point Cert, ASTM E126, E100-14 T1
Specific Gravity Range:
0.650 to 0.700 ASTM 82H-62
0.700 to 0.750 ASTM 103H-62
0.700 to 0.750 ASTM 83H-62
0.750 to 0.800 ASTM 104H-62
0.750 to 0.800 ASTM 84H-62
0.800 to 0.850 ASTM 105H-62
0.800 to 0.850 ASTM 85H-62
0.850 to 0.900 ASTM 86H-62
0.850 to 0.900 ASTM106H-62
0.900 to 0.950 ASTM 87H-62
0.950 to 1.00 ASTM 88H-62
Volume (Metric):
260 mL
275 mL
Unit Size:
Each
Product Code. 11819270
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Accuracy of hydrometer meets or exceeds ASTM™ E 100 specifications.
  • ASTM API 260mm ASTM Number: 104H 0.001 0.750 to 0.800 60°F Plain Form Hydrometer
  • W/3-pt certification
Order Info
A certificate of calibration is available; simply add the letter C to the end of the catalog number when ordering.
Compliance
Calibrated according to ASTM™ E 126 specifications
TRUSTED_SUSTAINABILITY
Specifications
Type Hydrometer
ASTM Number 104H
Certifications/Compliance 3-point Cert, ASTM E100-14 T1
For Use With (Application) Petroleum products
Specific Gravity Range 0.750 to 0.800 ASTM 104H-62
Volume (Metric) 260 mL
Temperature of Standardization 60°F