Learn More
Fisherbrand™ ASTM™ Specific Gravity 260mL Hydrometers for Liquids Heavier than Water
Product Code. 11767255 Shop All Fisherbrand Products
Click to view available options
ASTM Number:
125H
126H
127H
128H
129H
130H
131H
132H
Certifications/Compliance:
3-point Cert, ASTM E126, E100-14 T1
Specific Gravity Range:
1.00 to 1.05 ASTM 125H-62
1.05 to 1.10 ASTM 126H-62
1.10 to 1.15 ASTM 127H-62
1.15 to 1.20 ASTM 128H-62
1.20 to 1.25 ASTM 129H-62
1.25 to 1.30 ASTM 130H-62
1.30 to 1.35 ASTM 131H-62
1.35 to 1.40 ASTM 132H-62
Unit Size:
Each
Product Code. 11767255
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Accuracy meets or exceeds ASTM E 100 specifications.
  • Temperature of standardization: 60°F
  • Division: 0.001
  • Length: 260mm
Compliance
Calibrated according to ASTM™ E 126 specifications.
TRUSTED_SUSTAINABILITY
Specifications
ASTM Number 132H
Increments 0.001
Certifications/Compliance 3-point Cert, ASTM E126, E100-14 T1
Length (Metric) 260 mm
For Use With (Application) Liquids heavier than Water
Specific Gravity Range 1.35 to 1.40 ASTM 132H-62
Volume (Metric) 260 mL
Temperature of Standardization 60°F