Learn More
Fisherbrand™ Pouches for FB-GC10-1 Vertical Gel Caster
Product Code. 12444647
Click to view available options
Quantity:
100/Pk.
25/Pk.
Unit Size:
Pack of 100
Pack of 25
Product Code. 12444647
Brand: Fisherbrand™ GP425
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Pouches for use with FB-GC10-1 Vertical Gel Caster.
  • Dimensions: 10L × 10cmW
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Owl JGC-4 Gel Casting System
Type Gel Pouch
Quantity 25/Pk.
Length (Metric) 10 cm
Width (Metric) 10 cm