Learn More
Fisherbrand™ Combs for FB-VE16-1 Vertical Electrophoresis System
Product Code. 11983045
Click to view available options
Volume (Metric) Well:
51 μL
69 μL
96 μL
107 μL
129 μL
183 μL
201 μL
343 μL
Length (Metric) Footprint:
16 cm
20 cm
Teeth Count:
10
15
20
24
Width (Metric) Teeth:
0.29 cm
0.39 cm
0.61 cm
1.04 cm
Unit Size:
Each
Product Code. 11983045
Brand: Fisherbrand™ P120C
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Creates the wells in gels for electrophoresis
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Owl P1-CST Multiple Gradient Gel Caster
Type Comb
Gel Dimensions 16 x 14 cm
Teeth Count 20
Thickness (Metric) Teeth 0.08 cm
Volume (Metric) Well 69 μL
Length (English) Footprint 7.87 in.
Length (English) Gel 6.3 in.
Length (Metric) Footprint 20 cm
Length (Metric) Gel 16 cm
Width (English) Gel 5.5 in.
Width (English) Teeth 0.15 in.
Width (Metric) Footprint 13 cm
Width (Metric) Gel 14 cm
Width (Metric) Teeth 0.39 cm
Height (English) Footprint 7.08 in.
Height (Metric) Footprint 18 cm