Learn More
Fisherbrand™ 15-Tooth Combs for Owl™ P2-CST Multiple Gradient Gel Caster
Product Code. 11953055 Shop All Thermo Scientific Products
Change view
Click to view available options
Volume (Metric) Well:
144 μL
271 μL
Thickness (Metric) Teeth:
0.08 cm
0.15 cm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Well Thickness (Metric) Teeth unitSize
11953055 271 μL 0.15 cm Each
11943055 144 μL 0.08 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11953055 Supplier Fisherbrand™ Supplier No. P215D
Only null left
Add to basket
Add to basket
15-Tooth Combs for Owl™ P2-CST Multiple Gradient Gel Caster
  • For use with Owl P2-CST Multiple Gradient Gel Caster
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Owl P2-CST Multiple Gradient Gel Caster
Type Comb
Gel Dimensions 20 x 20 cm
Teeth Count 15
Thickness (Metric) Teeth 0.15 cm
Volume (Metric) Well 271 μL
Length (English) Footprint 9.84 in.
Length (English) Gel 7.8 in.
Length (Metric) Footprint 25 cm
Length (Metric) Gel 20 cm
Width (English) Gel 7.8 in.
Width (English) Teeth 0.32 in.
Width (Metric) Footprint 13 cm
Width (Metric) Gel 20 cm
Width (Metric) Teeth 0.82 cm
Height (English) Footprint 9.05 in.
Height (Metric) Footprint 23 cm