Learn More
Fisherbrand™ Wellwash™ and Wellwash™ Versa™ Microplate Strip Washer: Bottle Stands
Product Code. 11787996 Shop All Thermo Scientific Products
Change view
Click to view available options
Holds:
2 x 2 L Bottles
3 x 2 L Bottles, 1 x 4 L Bottle, Square
User Bottles
For Use With (Equipment):
Wellwash Microplate Strip Washer
Wellwash Microplate Strip Washer, Wellwash Versa Microplate Strip Washer
Wellwash Versa Microplate Strip Washer
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds For Use With (Equipment) Unit Size
11787996 2 x 2 L Bottles Wellwash Microplate Strip Washer Each
11728006 User Bottles Wellwash Microplate Strip Washer, Wellwash Versa Microplate Strip Washer Each
11738006 3 x 2 L Bottles, 1 x 4 L Bottle, Square Wellwash Versa Microplate Strip Washer Each
11748006 3 x 2 L Bottles, 1 x 4 L Bottle, Square Wellwash Microplate Strip Washer, Wellwash Versa Microplate Strip Washer Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11787996 Supplier Fisherbrand™ Supplier No. N10817
Estimated Shipment: 11-01-2027
Accessories for Fisher Scientific Wellwash and Wellwash Versa Microplate Strip Washer
For use with Fisher Scientific Wellwash and Wellwash Versa Microplate Strip Washer
ELISA-based assays, cell washing
TRUSTED_SUSTAINABILITY
Specifications
Holds 2 x 2 L Bottles
For Use With (Equipment) Wellwash Microplate Strip Washer
Product Type Bottle Stand