Learn More
Fisherbrand™ Probes for Model 50 and 120 Sonic Dismembrator
Image shows (left to right): 4423, 4422, 4435.
284.00€ - 373.00€
Product Code | Brand | Amplitude | Diameter (English) | Diameter (Metric) | Volume Range (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Amplitude | Diameter (English) | Diameter (Metric) | Volume Range (Metric) | Price | Quantity & Availability | ||
12921181
|
Fisherbrand™
|
200 μm maximum | 0.078 in. (5/64 in.) | 2 mm | 0.2 to 5 mL |
373.00€
Each |
Estimated Shipment: 14-05-2024
Log in to see stock. |
||
12911181
|
Fisherbrand™
|
180 μm maximum | 0.125 in. (1/8 in.) | 0.3 cm | 0.5 to 15 mL |
284.00€
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
||
12931181
|
Fisherbrand™
|
120 μm maximum | 0.25 in. | 0.63 cm | 10 to 50 mL |
325.00€
Each |
In Stock
Log in to see stock. |
||
Description
These Fisher Scientific™ Probes are designed for use with Model 50 and Model 120 Sonic Dismembrators.
- 5/64 in. Probe microtip for processing 0.2 to 5mL volumes
- 1/8 in. Probe microtip for processing 0.5 to 15mL volumes
- 1/4 in. Probe tip for processing 10 to 50mL volumes
Specifications
Probe | |
1 probe |