Learn More
Fisherbrand™ Precision Specific Gravity 325 - 368mm Long Hydrometers
Product Code. 11950364 Shop All Fisherbrand Products
Click to view available options
Certifications/Compliance:
NIST, ASTM E100, ASTM E126
Length (Metric):
325 mm
330 mm
335 mm
340 mm
345 mm
Specific Gravity Range:
0.640 to 0.710
0.700 to 0.770
0.760 to 0.830
0.820 to 0.890
0.880 to 0.950
0.940 to 1.010
1.000 to 1.070
1.060 to 1.130
1.120 to 1.190
1.240 to 1.310
1.300 to 1.370
1.360 to 1.430
1.420 to 1.490
1.48 to 1.55
Unit Size:
Each
Product Code. 11950364
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Requires 275mL liquid volume for submersion.
  • Use Cylinder 08-530K, available separately
  • Permanently marked with individual serial numbers
  • Accuracy meets or exceeds ASTM E 100 specifications
  • Calibrated according to ASTM E 126 specifications
  • With divisions of 0.001
  • Use Cylinder 08-530E, available separately
Order Info
A certificate of calibration is available; simply add the letter C to the end of the catalog number when ordering.
Certifications
A certificate is provided to indicate traceability to standards provided by NIST™.
TRUSTED_SUSTAINABILITY
Specifications
Increments 0.0005
Certifications/Compliance NIST, ASTM E100, ASTM E126
Length (Metric) 340 mm
For Use With (Equipment) 350mL Borosilicate Glass cylinder
Material Soda Lime Glass
For Use With (Application) Specific Gravity Measurement
Specific Gravity Range 0.640 to 0.710
Volume (Metric) 275 mL