Learn More
Fisherbrand™ Digital Thermometers with Stainless-Steel Stem, 0.375 in. LCD Screen and Protective Guard
Product Code. 17317659 Shop All Fisherbrand Products
Click to view available options
Accuracy:
±0.4°C
±1°C
Length (English):
3.75 in.
4.75 in.
5.5 in.
Unit Size:
Each
Product Code. 17317659
Brand: Fisherbrand™ 9876917
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Easy-to-read handheld digital thermometers
Easy-to-read Fisher Scientific™ Traceable Digital Thermometers with Stainless-Steel Stem are chemically resistant to acids, bases, solvents, and may be used to measure temperature of gases, food, and other semisolids.
  • Short-stem piercing models are ideal for carrying in a pocket
  • Longer probes are excellent for measuring sample temperatures while stirring in deep vessels or water baths
  • Thermometers with longer sampling rates allow thermometer to be removed from sample before reading—ideal when display is difficult to read in measuring position
  • For use in cuvets, soil, incubators, petroleum products and foods
Certifications
Individually serial-numbered Traceable Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™; indicates traceability to standards provided by NIST™.
Specifications
Certifications/Compliance ISO 17025, A2LA, NIST
Diameter (English) Probe 0.12 in.
For Use With (Application) Measuring temperature in soil, cuvettes, test tubes, food products, incubators and petroleum Hydrocarbons
Includes Protective case, traceable certificate and Silver Oxide battery
Length (English) Probe 4.75 in.
Length (Metric) 140 mm
Length (Metric) Probe 12 cm
Material Plastic
Resolution 0.1°C
Weight (English) 1.12 oz.
Weight (Metric) 31.7 g
Product Type Digital Thermometer
Length (English) 5.5 in.
Max. Temperature (English) 572°F
Max. Temperature (Metric) 300°C
Min. Temperature (English) -58°F
Min. Temperature (Metric) -50°C
Temperature Range (English) -58°F to +572°F
Temperature Range (Metric) -50°C to +300°C
Calibrated Yes
Accuracy ±1°C
Display Digital LCD
Width (Metric) 8 cm