Learn More
Electrothermal Capillary Tube for Digital Melting Point Apparatus
Product Code. 11748242
Change view
Click to view available options
Length (Metric):
1.5 mm
Unit Size:
Pack of 1000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
11748242 1.5 mm Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11748242 Supplier Electrothermal Supplier No. 0301067
  • ONLINE EXCLUSIVE PRICE
    260.00€
    181.52€ / Pack of 1000
    Save 78.48€
    30% Off
View all promotions
Estimated Shipment: 23-12-2026
For use with the IA9000 series Digital Melting Point Apparatus
Capillary tubes; 1.5-mm; 1000/pack. For use with the IA9000 series Digital Melting Point Apparatus
  • Capillary tubes
  • 1.5mm x 1000 tubes
Specifications
Product Type Capillary Tubes
Includes 1000 capillary tubes
Length (English) 0.05 in.
Length (Metric) 1.5 mm