Learn More
Fisherbrand™ Baume Hydrometers
Product Code. 11736933 Shop All Fisherbrand Products
Click to view available options
Certifications/Compliance:
Without 3-point certification
Increments:
0.1°
Length (Metric):
290 mm
295 mm
305 mm
310 mm
Baume Range:
19 to 31°
29 to 41°
40 to 51°
49 to 61°
60 to 70°
9 to 21°
Unit Size:
Each
Product Code. 11736933
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Baume heavy scale for working with liquids heavier than water.
Baume heavy scale for working with liquids heavier than water.
  • Permanently marked with individual serial numbers
Order Info
Use Fisherbrand cylinder 08-530J with 250mL of liquid for submersion, available separately
Certifications
A certificate is provided to indicate traceability to standards provided by NIST™.
Compliance
Accuracy meets or exceeds ASTM™ E 100 specifications, Calibrated according to ASTM E 126 specifications.
TRUSTED_SUSTAINABILITY
Specifications
Type Baume Hydrometer
Increments 0.1°
Certifications/Compliance Without 3-point certification
Length (Metric) 295 mm
Material Soda Lime Glass
Baume Range 49 to 61°
For Use With (Application) Baume 'Heavy' Scale For Liquids Heavier than Water