Learn More
Fisherbrand™ Air-Powered Analog Overhead Stirrers
Product Code. 15112997 Shop All Fisherbrand Products
Click to view available options
Speed:
10,000 rpm
1000 rpm
1200 rpm
3000 rpm
Capacity:
10 L
20 L
50 L
Torque:
45
203
294
633
Unit Size:
Each
Product Code. 15112997
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Includes
  • A 9/16 in. dia. x 10 in. L support rod
  • Four ft. air hose with double fittings for connection to air supply
    • Variable speed and explosion-proof stirrers ideal for safely mixing all types of solvents, lacquers and volatile chemicals without the possiblity of spark ignition.
      • Needle valve allows variation in mixing speed
      • Special muffler ensures quieter operation
      • Reversible models have motors which can be set to rotate in either direction by connecting the air supply line to the right port for counter-clockwise rotation or the left port for clockwise rotation
      • Can be used with flammable chemicals for explosion proof mixing
      Order Info
      Shaft, propeller, stand and clamp available separately
Specifications
Speed 1200 rpm
Capacity 20 L
Display Analog
Voltage 0/1 V
Diameter (English) 0.37 in.
Diameter (Metric) Shaft 0.95 cm
Height (English) 10 in.
Height (Metric) 25 cm
Length (English) 12 in.
Length (Metric) 30 cm
Horsepower 0.3
For Use With (Application) Safely mixing all types of solvents, lacquers and volatile chemicals
Includes 10 L x 0.56 in. dia. support rod, 4 ft. air hose with double fittings for connection to air supply
Material Steel Aluminum
Pressure (English) Air Supply 30 to 80 psi
Torque 203
Type Air Powered Overhead Stirrer
Viscosity 10,000 mPas
Weight (English) 6 lb.
Weight (Metric) 2.7 kg