Learn More
Fisherbrand™ accumet™ Membrane Caps for Gas-Sensing Combination ISEs
Product Code. 11530134 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Diameter (Metric):
12 mm
Unit Size:
Pack of 3
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
11530134 12 mm Pack of 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11530134
  • ONLINE EXCLUSIVE PRICE
    211.00€
    155.91€ / Pack of 3
    Save 55.09€
    26% Off
Estimated Shipment: 07-12-2026
Set of three replacement membrane caps with O-rings.
  • For Ammonia Gas-Sensing Combination ISEs 13-620-508, 13-620-509; Carbon Dioxide Gas-Sensing Combination ISEs 13-620-510, 13-620-511
  • Set of three replacement membrane caps
  • With O-rings
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Ammonia Gas-sensing Combination ISE
Type Membrane Cap
Diameter (Metric) 12 mm