Learn More
Fisher BioReagents™ Nucleotides
Product Code. 10658973
Click to view available options
Product Type:
dATP
dCTP
dGTP
dNTP Set
Quantity:
100 μL
400 μL
Unit Size:
1 set
400µL
Product Code. 10658973
Brand: Fisher BioReagents™ BP25604
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Nucleotides for PCR with a purity level of >98% triphosphate
  • Functionally tested in PCR
  • Can be used in dideoxy sequencing, mutagenesis, and cDNA synthesis applications
  • DNase-, RNase-, and protease-free
  • PCR dNTP Mix is a solution of dATP, dCTP, dGTP, and dTTP
  • dNTP Set contains one tube each of dATP, dCTP, dGTP, and dTTP (4 tubes/set)
TRUSTED_SUSTAINABILITY
Specifications
Format Liquid
pH 7 to 7.5
Molecular Weight (g/mol) 579.2
Product Type dATP
Concentration 100 mM
Purity >98%
Quantity 400 μL
Color Colorless
For Use With (Application) Dideoxy sequencing, mutagenesis and cDNA synthesis