Learn More
Thermo Scientific™ Finntip™ Universal Pipette Tips
Product Code. 10053860
Click to view available options
Description:
Finntip 250 Universal refill starter kit, Non-Filtered, Non-sterile, Refill
Finntip 250 Universal, Non-Filtered, Non-sterile, Box
Finntip 250 Universal, Non-Filtered, Non-sterile, Bulk
Volume (Metric):
250 μL
Format:
Box
Bulk
Refill
Sterility:
Non-sterile
Unit Size:
1 set
Pack of 20000
Pack of 500
Product Code. 10053860
Brand: Thermo Scientific™ 9400250
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Ensure accurate, precise pipetting with these universal pipette tips, designed using our knowledge of liquid handling, with uniformly smooth, hydrophobic surfaces that prevent liquid retention.
Ensure accurate, precise pipetting with Thermo Scientific™ Finntip™ Universal Pipette Tips, manufactured using high quality raw materials and the latest molding techniques. Finntips are designed using our knowledge of liquid handling. Our modern automated production facilities, and our expertise, enable us to produce uniformly smooth, hydrophobic surfaces on the inside and outside of our tips to prevent liquid retention.
Features:
  • Smooth, uniform surface prevents liquid retention
  • Volume range from 0.2μL to 10mL
  • Available sterile or nonsterile
  • Packaging options include racks, bags, boxes, bulk and refill packs
  • Color-coded racks to match the compatible Finnpipette
  • Color-coding only applies to rack and refill packaging
Note:
Manufactured according to ISO 9001, ISO 14001 and ISO 13485 (CE/IVD certified).
Limited warranty period:
Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warrantied from the date we ship the product and for ninety (90) days thereafter.
TRUSTED_SUSTAINABILITY
Specifications
Description Finntip 250 Universal, Non-Filtered, Non-sterile, Box
Volume (Metric) 250 μL
Filtered Non-filtered
Sterility Non-sterile
Color Yellow
Model Finntip 250 Universal
Length (English) 2 in.
Length (Metric) 5.2cm
Certifications/Compliance ISO 9001, ISO 14001, ISO 13485
Format Box
Tip Style Universal Fit
Packaging 500 tips
For Use With (Equipment) Thermo Scientific Finnpipette 250
Product Line FinnTip