Learn More
Cole-Parmer™ Lab Markers
Product Code. 11700239 Shop All Cole-Parmer Products
Click to view available options
Color:
Blue
Red
Unit Size:
Pack of 10
Product Code. 11700239
Brand: Cole-Parmer™ 0996410
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Write on a variety of surfaces with these versatile lab markers. Cole-Parmer™ Lab Markers permanently mark plastic; wash off glass, metal, or porcelain. Markers won’t bleed or smudge when subjected to water or alcohol.
  • Fine-point fiber tip
  • Quick-drying ink
  • Writes on cold, wet surfaces
  • Available in four different colors
Specifications
Color Blue
For Use With (Application) For Glass, Porcelain or Metal surfaces
Quantity 10/Pk.