Learn More
Brady™ Fine Pen
Product Code. 16318186 Shop All Brady Products
Change view
Click to view available options
Color:
Black
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
16318186 Black Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16318186 Supplier Brady™ Supplier No. 335091
  • ONLINE EXCLUSIVE PRICE
    30.20€
    24.85€ / Pack of 5
    Save 5.35€
    17% Off
View all promotions
In Stock
Use this labeling pen on a variety of surfaces. Brady™ Fine Pens use a quick drying, permanent ink.
Specially formulated to resist oil, abrasion and fading.
Specifications
Tip Style Fine
Alternate Part Number BFS-05
Product Type Permanent Marker
Color Black
Description Pen - Fine
For Use With (Application) Used For Marking Laboratory Glass Wares
Quantity 1 Pk.