Learn More
Merck Filtration Kit
2380.00€ - 2755.00€
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
11770403
|
Merck
KITFILTFG |
FGLP |
2590.00€
1 set |
Estimated Shipment: 23-07-2024
Log in to see stock. |
|||||
11750413
|
Merck
KITFILTLC |
FHLC |
2755.00€
1 set |
Estimated Shipment: 23-07-2024
Log in to see stock. |
|||||
11780403
|
Merck
KITFILTFH |
FHLP |
2590.00€
1 set |
Estimated Shipment: 23-07-2024
Log in to see stock. |
|||||
11790403
|
Merck
KITFILTGN |
GNWP |
2450.00€
1 set |
Estimated Shipment: 23-07-2024
Log in to see stock. |
|||||
11700413
|
Merck
KITFILTGS |
GSWP |
2415.00€
1 set |
Estimated Shipment: 23-07-2024
Log in to see stock. |
|||||
11710413
|
Merck
KITFILTGV |
GVWP |
2545.00€
1 set |
Estimated Shipment: 23-07-2024
Log in to see stock. |
|||||
11720413
|
Merck
KITFILTHA |
HAWP |
2380.00€
1 set |
Estimated Shipment: 23-07-2024
Log in to see stock. |
|||||
11740413
|
Merck
KITFILTHV |
HVLP |
2545.00€
1 set |
Estimated Shipment: 23-07-2024
Log in to see stock. |
|||||
11730413
|
Merck
KITFILTHN |
HNWP |
2450.00€
1 set |
Estimated Shipment: 23-07-2024
Log in to see stock. |
|||||