Learn More
Syngene™ Short Pass Emission Filter for 3 or 7 Position Selector Accessory
GREENER_CHOICE
Product Code. 12874078 Shop All Syngene Products
Click to view available options
Wavelength:
515-599nm
Unit Size:
Each
Product Code. 12874078
Brand: Syngene™ FILTSPGX
Only null left
Add to basket
Add to basket
Syngene™ Short Pass Emission Filter for 3 or 7 Position Selector Accessory works in conjunction with the Syngene Gbox and Gbox Mini Systems.
The G:BOX iChemi series of gel documentation systems offers a range of camera, lighting and filter options to cover all applications.
The innovative design gives the user an incredible array of configuration choices. There are 4 basic configurations of CCD camera. This ensures that there is a G:BOX iChemi to suit all budgets and all applications. The new G:BOX iChemi systems intelligently assess a sample and then automatically set the optimum exposure time and sensitivity settings to obtain the very best image possible. Further improvements to the scaling of the samples also ensure optimum display settings giving images of superb and outstanding quality. In summary, the G:BOX iChemi is the perfect system with performance to match. Choose from the four basic configurations and simply add any options you need.
  • G:BOX iChemi: High resolution version for general purpose fluorescence and chemiluminescence
  • G:BOX iChemi XR: Super sensitive low noise systems for high performance applications
  • G:BOX iChemi XL: Mega pixel system with superior capture speed
  • G:BOX iChemi XT: Mega pixel system with superior performance
Gel documentation and analysis
  • Modular system
  • Choice of camera - the right camera for your application
  • Light-tight darkroom - suitable for fluorescence and chemiluminescence
  • 16 bit camera (EF) - very accurate gel data
  • Auto focus configuration and focus indicator - precise focusing every time
  • Auto exposure and series capture - easy image capture
  • Integration control - capture faint bands or features from any gel
  • Neutral fielding (option) - perfect background correction
  • Filter selection options - can detect a wide range of stains and dyes
  • Effective pixels - extended resolution
  • GeneTools - analysis software
Scan and document electrophoresis gels stained with:
  • Ethidium™ Bromide, Coomassie™ Blue, Silver Stain, SYBR™ Gold, SYBR™ Green, SYBR™ Safe, GelStar™, SYPRO™ Red , SYPRO™ Ruby, SYPRO™ Orange, Fluorescein, Rhodamine Red™ , Texas Red™, Pro-Q™ Diamond , Deep Purple™ , Q-dots, as well as:
  • Chemiluminescence - most common substrates including: ECL™, SuperSignal™, LumiGlo™, West Dura™
  • Agar plates
  • Cells in flasks
  • Autoradiographs
  • Membranes
  • Spots and slot blots
  • TLC plates
  • Microtitre plates
  • Macro-arrays
Specifications
For Use With (Equipment) Gbox and Gbox Mini
Wavelength 515-599nm