Learn More
Brand™ Filling tube Seripettor with O ring polypropylene 250mm Brand
Shop All Brand Products
Click to view available options
For Use With (Equipment):
Seripettor™ Bottletop Dispenser
Unit Size:
Each
Specifications
Specifications
| Description | Filling tube for Seripettor |
| For Use With (Equipment) | Seripettor™ Bottletop Dispenser |
| Length (Metric) | 250 mm |