Learn More
Ohaus™ Field Test Scale
Product Code. 10531222 Shop All Ohaus Products
Change view
Click to view available options
Metric Capacity: 16000g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10531222 Metric Capacity: 16000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10531222 Supplier Ohaus™ Supplier No. 80000008
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
The Field Test Scale provides accurate weighing of a wide variety of heavy materials, even under the most adverse conditions. The Field Test Scale is ruggedly built and designed for field applications where accurate measurements are essential
TRUSTED_SUSTAINABILITY
Specifications
Depth (English) 15.35 in.
Depth (Metric) 390.00 mm
Height (English) 8.22 in.
Height (Metric) 209.000 mm
Includes Epoxy coated steel pan
Model 2400-11
Material Metal Base and Beam, Oversized Stainless Steel Pan
Width (English) 7.64 in.
Width (Metric) 194.00 mm
For Use With (Application) In a Laboratory, in an Intrinsically Safe Environm
Product Line Field Test
Readability 5.000000 g
Description Mechanical Scale, 2400-11
Languages Chinese, English (US), French, German, Italian, Japanese, Korean, Polish, Portuguese (BR), Spanish (MX)
Display Type Sliding Poise and Graduated Beam
Type Field Test Scale