missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Fibrinogen Native Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Specifications
Specifications
| Concentration | 1.25 mg/mL |
| For Use With (Application) | ELISA, Functional Assay |
| Formulation | Citrate buffered saline |
| Gene ID (Entrez) | 2243 |
| Molecular Weight (g/mol) | MolecularWeight-theroretical: 340 kDa |
| Quantity | 25 mg |
| Storage Requirements | Store at -20°C. |
| Gene Alias | Fib2, fibrinogen alpha chain, fibrinogen, A alpha polypeptide, MGC119422, MGC119423, MGC119425 |
| Gene Symbol | FGA |
| Research Category | Angiogenesis, Cancer, Cell Cycle and Replication |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction