Learn More
Thermo Scientific™ Fiberlite™ H3-LV Large Volume Swinging Bucket Rotor
Description
Conduct high-volume microplate processing with the Thermo Scientific™ Fiberlite™ H3-LV Large Volume Swinging Bucket Rotor.
Features:
Fiberlite rotors:
- Lightweight carbon fiber rotors reduce run-times by achieving higher G-forces
- Corrosion-and fatigue-resistant design ensures structural integrity
- Rotors are backed by a 15-year warranty and are compatible with Thermo Scientific™ Nalgene™ bottles and tubes for complete centrifuge solutions
Recommended for:
Pelleting cells and cellular debris, protein precipitation, plasmid purification and collecting physiological fluids for diagnostic tests
Note:
†Performance may vary by centrifuge model.
Specifications
Specifications
| Description | Fiberlite H3-LV Large Volume Swinging Bucket Rotor for Thermo Scientific Sorvall Legend XT, Multifuge X3 Series |
| Type | Swinging Bucket Rotor |
| Capacity | 28 Standard or 8 Deepwell Plates |
| Max. RCF | 1,840 x g |
| Max. Speed | 3,600 rpm |
| For Use With (Equipment) | Thermo Scientific Sorvall Legend XT/XF, Multifuge X3/X3F and SL 40F Centrifuges |
| Angle | 90° |
| Biocontainment | Yes |