Learn More
Thermo Scientific™ Fiberlite™ F14-6 x 250LE Rotor Adapters
Product Code. p-4527810 Shop All Thermo Scientific Products
Click to view available options
Capacity:
15 mL Conical Tube (Set of 2)
50 mL Conical Tube (Set of 2)
Unit Size:
Pack of 2
Product Code. 11898362
Brand: Thermo Scientific™ 75100136
Combine this rotor Adapter with the Fiberlite F14-6 x 250LE fixed-angle rotor.
Combine Thermo Scientific™ Fiberlite™ F14-6 x 250LE Rotor Adapters with the Fiberlite F14-6 x 250LE fixed-angle rotor.
Note:
  • Adapters 00369 and 00456 must be used together for 7 mL round-bottom tubes
Ordering Information:
Fiberlite F14-6x250LE fixed-angle rotor body (75003662) is also available. Contact your sales representative for more information.
TRUSTED_SUSTAINABILITY
Specifications
Type Rotor Adapter
Tube Type Conical
Capacity 50 mL Conical Tube (Set of 2)
Tube Size 50 mL