Learn More
Brady™ Fiberglass-Reinforced Nylon Miniature Locking System for Circuit Breakers
Shop All Brady ProductsDescription
- Requires no tools to install
- Lockout easily installs with the aid of a push button
- Tie-bar style lockout has a thumb wheel for quick application
Specifications
Specifications
| Product Type | Miniature Locking System for Circuit Breakers |
| Alternate Part Number | ELEC. CIRCUIT BREAKER L/O EURO POS. PK6 |
| Color | Red/Black |
| Material | Fiberglass-reinforced Nylon |
| Certifications/Compliance | UL 94 HB |
| Description | Pin-Out Standard |
| Diameter (English) | 0.5 in. |
| Diameter (Metric) | 24.13 mm |
| For Use With (Application) | Lockout Tagout |
| Height (English) | 1.638 in. |
| Show More |
Product Title
Vous avez repéré une opportunité d'amélioration ?