Learn More
SCHOTT AG Lighting and Imaging™ Fiber Optic Combi Light Guide
Product Code. 15743032 Shop All Schott Products
Change view
Click to view available options
For Use With (Equipment):
Cold Light Source KL1600LED, KL2500LED or KL1500HAL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15743032 Cold Light Source KL1600LED, KL2500LED or KL1500HAL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15743032 Supplier SCHOTT AG Lighting and Imaging™ Supplier No. 170402
Estimated Shipment: 03-12-2026
Fiber optic flex and stay double light guide in a black lacquered semi-rigid gooseneck tubing with an additional 800 mm fiber optic flexible light guide in a black metal-PVC jacketing.
  • Light guide with eco-friendly Puravis fiber optics.
  • Precise positioning offers complete control of light placement.
  • The combination of a double gooseneck light guide 4.5 mm dia. x 500 mm and a flexible light guide 4.5 mm dia. x 800 mm allows it to move further from the lightsource.
  • Active Fiber optic bundle: 2 x 4.5 mm for Gooseneck Outer Jacketing, 11 mm for gooseneck, and 14 mm for flexible.
  • Can be stationary fixed at the microscope post with the adapter 158 402.
  • To use with Cold light source KL1500HAL, KL1600LED or KL2500LED.
Specifications
Type Fiber Optic Lightguide
Diameter (Metric) 11 mm (Gooseneck), 14 mm (Flexible)
For Use With (Application) Stereo Microscopy Illumination
For Use With (Equipment) Cold Light Source KL1600LED, KL2500LED or KL1500HAL
Length (Metric) 2 x 500 mm for gooseneck and 800 mm for flexible
Material Eco-friendly Puravis Glass Optical Fibers